missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM29 Monoclonal Antibody (OTI5C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812274
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype.Specifications
| TRIM29 | |
| Monoclonal | |
| Unconjugated | |
| TRIM29 | |
| ATDC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23650 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C11 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14134 | |
| TRIM29 | |
| Human recombit protein fragment corresponding to amino acids 1-352 of human TRIM29 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction