missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM30 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41087H
This item is not returnable.
View return policy
Description
TRIM30 Polyclonal antibody specifically detects TRIM30 in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| TRIM30 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Rpt1, Rpt-1, Trim30a | |
| Antibody was raised against a 21 amino acid synthetic peptide near the amino terminus of the mouse TRIM30. The immunogen is located within amino acids 110 - 160 of TRIM30. Amino Acid Squence: RSQEHRGHQTALIEEVDQEYK | |
| 0.1 mL | |
| Innate Immunity | |
| 20128 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| HRP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction