missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569224
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
Specifications
| TRIM36 | |
| Polyclonal | |
| Unconjugated | |
| TRIM36 | |
| Acrosome RBCC protein; D18Wsu100e; E3 ubiquitin-protein ligase Trim36; haprin; RBCC728; RING finger protein 98; RING-type E3 ubiquitin transferase TRIM36; RNF98; trim36; tripartite motif containing 36; tripartite motif protein 36; tripartite motif-containing 36; tripartite motif-containing protein 36; wu:fj48f01; zgc:153447; zgc:63476; Zinc-binding protein Rbcc728 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 291597 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| TRIM36 | |
| synthetic peptide directed towards the middle region of human TRIM36. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction