missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM37 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-80147
This item is not returnable.
View return policy
Description
TRIM37 Polyclonal specifically detects TRIM37 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| TRIM37 | |
| Polyclonal | |
| Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA 1:64000 | |
| EC 6.3.2.-, KIAA0898Tripartite motif-containing protein 37, Mulibrey nanism protein, MULRING-B-box-coiled-coil protein, POB1E3 ubiquitin-protein ligase TRIM37, TEF3, tripartite motif containing 37, tripartite motif-containing 37 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reported variants represent identical protein (NP_056109.1; NP_001005207.1). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| TRIM37 | |
| Peptide with sequence C-EDLSFNTDENSGR corresponding to C-Terminus according to NP_056109.1. | |
| 0.1 mg | |
| Stem Cell Markers | |
| 4591 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction