missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM37 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110519

Product Code. 17168443

  • € 502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LGMELLAKSS VANCYIRNST NKKSNSPKPA RSSVAGSLSL RRAVDPGENS RSKGDCQTLS EGSPGSSQSG SRHSSPRALI HGSIGDILPK TEDRQCKALD SDAVVVAVFS GLPAVEKRRK MVTLGANAKG GHLEGLQMTD LENNSETGEL QPVLPEGASA APEEGMSSDS DIECDTENEE QEEHTSVGGF HDSFMVMTQP PDEDTHSSFP DGEQIGPEDL SFNTDENSGR.

Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIM37
Polyclonal
Unconjugated
TRIM37
1110032A10Rik; 2810004E07Rik; AI848587; AU043018; E3 ubiquitin-protein ligase TRIM37; Kiaa0898; mKIAA0898; MUL; mulibrey nanism protein; POB1; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TEF3; TRAF encompassing factor 3; Trim37; tripartite motif containing 37; tripartite motif protein 37; tripartite motif-containing 37; tripartite motif-containing protein 37
Rabbit
Affinity Chromatography
RUO
360592, 68729
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.24 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q6PCX9
TRIM37
Recombinant fusion protein containing a sequence corresponding to amino acids 735-964 of human TRIM37 (NP_0010052071).
100 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.