missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM37 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17808902
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17808902 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17808902 Supplier Invitrogen™ Supplier No. PA5142017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: Expected to recognise all reported isoforms a to i. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM37
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TRIM37
Gene Accession No. O94972
Gene Alias 1110032A10Rik; 2810004E07Rik; AI848587; AU043018; E3 ubiquitin-protein ligase TRIM37; Kiaa0898; mKIAA0898; MUL; mulibrey nanism protein; POB1; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TEF3; TRAF encompassing factor 3; Trim37; tripartite motif containing 37; tripartite motif protein 37; tripartite motif-containing 37; tripartite motif-containing protein 37
Gene Symbols TRIM37
Host Species Goat
Immunogen Peptide with sequence C-EDLSFNTDENSGR.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4591
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.