missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM38 Monoclonal Antibody (OTI113D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505981
This item is not returnable.
View return policy
Description
This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.Specifications
| TRIM38 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00635 | |
| TRIM38 | |
| Human recombit protein fragment corresponding to amino acids 1-265 of human TRIM38 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI113D3 | |
| Unconjugated | |
| TRIM38 | |
| RNF15, RORET | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10475 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction