missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM38 Monoclonal Antibody (OTI2G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505969S
This item is not returnable.
View return policy
Description
TRIM38 Monoclonal antibody specifically detects TRIM38 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| TRIM38 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78358 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1485 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G9 | |
| Unconjugated | |
| CTAG1B | |
| E3 ubiquitin-protein ligase TRIM38; Gm23; hypothetical protein LOC531022; MGC8946; RING finger protein 15; RING-type E3 ubiquitin transferase TRIM38; RNF15; Ro/SSA ribonucleoprotein homolog; RORET; TRIM38; tripartite motif containing 38; tripartite motif-containing 38; tripartite motif-containing protein 38; tripartite motif-containing protein 38; E3 ubiquitin-protein ligase TRIM38; tripartite motif-containing protein 38; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM38; zinc finger protein RoRet | |
| Full length human recombinant protein of human CTAG1B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction