missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM38 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538516
This item is not returnable.
View return policy
Description
This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4. [provided by RefSeq, Jul 2012].
Specifications
| TRIM38 | |
| Polyclonal | |
| Unconjugated | |
| TRIM38 | |
| RNF15, RORET | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10475, 214158 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00635 | |
| TRIM38 | |
| A synthetic peptide derived from the internal region of human TRIM38 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction