missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM41 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 460.00
Specifications
| Antigen | TRIM41 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2 μg/ml, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/ Immunofluorescence 5 - 10 μg/ml, Immunohistochemistry-Paraffin 1:100 - 1:300 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643198
|
Novus Biologicals
NBP3-05293 |
0.1 mg |
€ 460.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601547
|
Novus Biologicals
NBP3-05293SS |
0.025 mg |
€ 188.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM41 Polyclonal antibody specifically detects TRIM41 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| TRIM41 | |
| Western Blot 1-2 μg/ml, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/ Immunofluorescence 5 - 10 μg/ml, Immunohistochemistry-Paraffin 1:100 - 1:300 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| tripartite motif containing 41 | |
| Partial synthetic peptide made to an internal portion of the human TRIM41 protein (between amino acids 400-470) [UniProt Q8WV44]. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy | |
| PBS | |
| 90933 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title