missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555013
This item is not returnable.
View return policy
Description
Immunogen sequence: RESTHHKEKV GPGGSSVGSG DASSSRHHHR RRRLHLPQQP LLQREVWCVG TNGKRYQAQS STEQTLLSPS EKPRRFGVYL DYEA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 ). See TRIM45.
Specifications
| TRIM41 | |
| Polyclonal | |
| Unconjugated | |
| TRIM41 | |
| AW552703; BC020156; E3 ubiquitin-protein ligase TRIM41; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM41; R75223; RINCK; RING finger-interacting protein with C kinase; RING-finger protein that interacts with C kinase; RING-type E3 ubiquitin transferase TRIM41; Trim41; tripartite motif containing 41; tripartite motif protein 41; tripartite motif-containing 41; tripartite motif-containing protein 41 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WV44 | |
| TRIM41 | |
| Recombinant protein corresponding to Human TRIM41. Recombinant protein control fragment (Product #RP-93582). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction