missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM42 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA531893

Product Code. 13426417

  • € 526.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: U87-MG. Predicted reactivity: Human (99%), Mouse (88%), Rat (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, namely a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIM42
Polyclonal
Unconjugated
TRIM42
PPP1R40; protein phosphatase 1, regulatory subunit 40; T4A1; TRIM42; tripartite motif containing 42; tripartite motif-containing 42; tripartite motif-containing protein 42
Rabbit
Antigen affinity chromatography
RUO
287015
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.58 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q8IWZ5
TRIM42
Recombinant protein encompassing a sequence within the center region of human TRIM42. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.