missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM43 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20719
This item is not returnable.
View return policy
Description
TRIM43 Polyclonal specifically detects TRIM43 in Human samples. It is validated for Western Blot.
Specifications
| TRIM43 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| tripartite motif containing 43, tripartite motif-containing 43, tripartite motif-containing protein 43 | |
| Rabbit | |
| 52 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96BQ3 | |
| TRIM43 | |
| Recombinant protein encompassing a sequence within the center region of human TRIM43. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 129868 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction