missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM44 Antibody (OTI1E10), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74611JF646
This item is not returnable.
View return policy
Description
TRIM44 Monoclonal specifically detects TRIM44 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TRIM44 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRIM44 | |
| Full length recombinant protein of human TRIM44 (NP_060053) produced in HEK293T cells. | |
| 0.1 mL | |
| 54765.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DIPBMGC3490, HSA249128, MC7, Protein DIPB, tripartite motif containing 44, tripartite motif-containing 44, tripartite motif-containing protein 44 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction