Learn More
TRIM44 Monoclonal Antibody (OTI2B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505513
Encompass_Preferred
 View more versions of this product
Product Code. 17584072
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.
Specifications
Specifications
TRIM44
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96DX7
TRIM44
Full length human recombit protein of human TRIM44 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2B9
Unconjugated
TRIM44
AN3, DIPB, HSA249128, MC7
Mouse
Affinity Chromatography
RUO
54765
-20° C, Avoid Freeze/Thaw Cycles