missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM44 Monoclonal Antibody (OTI5C2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526157
This item is not returnable.
View return policy
Description
TRIM44 Monoclonal Antibody for Western Blot, IHC (P)
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.
Specifications
| TRIM44 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96DX7 | |
| TRIM44 | |
| Full length human recombinant protein of TRIM44 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C2 | |
| Unconjugated | |
| TRIM44 | |
| brain cDNA 7; DIPB; DIPB protein; HSA249128; Mc7; Protein DIPB; Protein Mc7; Trim44; tripartite motif containing 44; tripartite motif protein 44; tripartite motif-containing 44; tripartite motif-containing protein 44 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54765 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido