Learn More
TRIM44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11511-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against full length of human TRIM44.
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.Specifications
| TRIM44 | |
| Polyclonal | |
| Unconjugated | |
| TRIM44 | |
| DIPB, HSA249128, MC7, Protein DIPB, TRIM44, tripartite motif containing 44 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362172, 54765, 80985 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6QA27, Q96DX7, Q9QXA7 | |
| Trim44 | |
| TRIM44 Fusion Protein Ag2070 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |