missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM45 Antibody (OTI3G3), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74612UV
This item is not returnable.
View return policy
Description
TRIM45 Monoclonal specifically detects TRIM45 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TRIM45 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRIM45 | |
| Recombinant protein fragment corresponding to amino acids 288-580 of human TRIM45 (NP_079464) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3G3 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| RING finger protein 99, RNF99FLJ13181, tripartite motif containing 45, tripartite motif-containing 45, tripartite motif-containing protein 45 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Zinc Finger | |
| 80263.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction