missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM45 Antibody (OTI3G3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46223
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Immunohistochemistry, Immunofluorescence
TRIM45 Monoclonal specifically detects TRIM45 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence.
Specifications
| TRIM45 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
| RING finger protein 99, RNF99FLJ13181, tripartite motif containing 45, tripartite motif-containing 45, tripartite motif-containing protein 45 | |
| Mouse | |
| 64.2 kDa | |
| 0.1 mL | |
| Apoptosis, Zinc Finger | |
| 80263 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunofluorescence | |
| OTI3G3 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| TRIM45 | |
| Human recombinant protein fragment corresponding to amino acids 288-580 of human TRIM45(NP_079464) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction