missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM45 Monoclonal Antibody (OTI3G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505920S
This item is not returnable.
View return policy
Description
TRIM45 Monoclonal antibody specifically detects TRIM45 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)Specifications
| TRIM45 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H8W5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80263 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| OTI3G3 | |
| Unconjugated | |
| TRIM45 | |
| 4921530N01Rik; FLJ13181; RING finger protein 99; RNF99; TRIM45; tripartite motif containing 45; tripartite motif protein 45; tripartite motif-containing 45; tripartite motif-containing protein 45 | |
| Human recombinant protein fragment corresponding to amino acids 288-580 of human TRIM45 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction