missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM45 Monoclonal Antibody (OTI3G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505920
This item is not returnable.
View return policy
Description
TRIM45 is a member of the tripartite motif family. It may function as a transcriptional repressor of the mitogen-activated protein kinase pathway. Alternatively spliced transcript variants have been described. TRIM45 belongs to a family of tripartite motif (TRIM) proteins that play important roles in a variety of cellular functions, including cell proliferation, differentiation, development, oncogenesis, and apoptosis.Specifications
| TRIM45 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H8W5 | |
| TRIM45 | |
| Human recombit protein fragment corresponding to amino acids 288-580 of human TRIM45 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI3G3 | |
| Unconjugated | |
| TRIM45 | |
| RNF99 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80263 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction