missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM45 Monoclonal Antibody (OTI4F1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526229
This item is not returnable.
View return policy
Description
TRIM45 Monoclonal Antibody for Western Blot, IHC (P)
TRIM45 is a member of the tripartite motif family. It may function as a transcriptional repressor of the mitogen-activated protein kinase pathway. Alternatively spliced transcript variants have been described. TRIM45 belongs to a family of tripartite motif (TRIM) proteins that play important roles in a variety of cellular functions, including cell proliferation, differentiation, development, oncogenesis, and apoptosis.
Specifications
| TRIM45 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H8W5 | |
| TRIM45 | |
| Human recombinant protein fragment corresponding to amino acids 288-580 of TRIM45 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F1 | |
| Unconjugated | |
| TRIM45 | |
| 4921530N01Rik; FLJ13181; RING finger protein 99; RNF99; TRIM45; tripartite motif containing 45; tripartite motif protein 45; tripartite motif-containing 45; tripartite motif-containing protein 45 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80263 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction