Learn More
TRIM45 Mouse anti-Human, Clone: OTI4F1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505922
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TRIM45 is a member of the tripartite motif family. It may function as a transcriptional repressor of the mitogen-activated protein kinase pathway. Alternatively spliced transcript variants have been described. TRIM45 belongs to a family of tripartite motif (TRIM) proteins that play important roles in a variety of cellular functions, including cell proliferation, differentiation, development, oncogenesis, and apoptosis.Specifications
| TRIM45 | |
| Monoclonal | |
| Unconjugated | |
| TRIM45 | |
| RNF99 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80263 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H8W5 | |
| TRIM45 | |
| Human recombit protein fragment corresponding to amino acids 288-580 of human TRIM45 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.