missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM47 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | TRIM47 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226790
|
Novus Biologicals
NBP3-35790-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229428
|
Novus Biologicals
NBP3-35790-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM47 Polyclonal antibody specifically detects TRIM47 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| TRIM47 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 91107 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Gene overexpressed in astrocytoma protein, GOARING finger protein 100, RNF100tripartite motif-containing protein 47, tripartite motif containing 47, tripartite motif-containing 47 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 356-530 of human TRIM47 (NP_258411.2).,, Sequence:, ELSFTKSSQAVRAVRDMLAVACVNQWEQLRGPGGNEDGPQKLDSEADAEPQDLESTNLLESEAPRDYFLKFAYIVDLDSDTADKFLQLFGTKGVKRVLCPINYPLSPTRFTHCEQVLGEGALDRGTYYWEVEIIEGWVSMGVMAEDFSPQEPYDRGRLGRNAHSCCLQWNGRSFS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title