missing translation for 'onlineSavingsMsg'
Learn More

TRIM73 Polyclonal Antibody
GREENER_CHOICE

TRIM50 Polyclonal Antibody

Brand:  Invitrogen PA565481

Product Code. 15968224

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRIM50 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIM73
Polyclonal
Unconjugated
TRIM73
TRIM50B; TRIM73; tripartite motif containing 73; tripartite motif-containing protein 50B; Tripartite motif-containing protein 73
Rabbit
Antigen Affinity Chromatography
RUO
375593
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q86UV7
TRIM73
Recombinant Human TRIM50. Recombinant protein control fragment (Product #RP-105617).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.