missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM54 Monoclonal Antibody (OTI6F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803765S
This item is not returnable.
View return policy
Description
TRIM54 Monoclonal antibody specifically detects TRIM54 in Human samples. It is validated for Western BlotSpecifications
| TRIM54 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5DX21 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 152404 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6F5 | |
| Unconjugated | |
| Igsf11 | |
| 4930486E09Rik; 4930566I02Rik; MURF; MURF3; MuRF-3; Muscle-specific RING finger protein; Muscle-specific RING finger protein 3; muscle-specific RING-finger protein; muscle-specific RING-finger protein 3; OTTHUMP00000122598; ring finger protein 30; Rnf30; TRIM54; tripartite motif containing 54; tripartite motif-containing 54; tripartite motif-containing protein 54 | |
| Human recombinant protein fragment corresponding to amino acids 23-241 of human IGSF11 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction