missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM56 Antibody (S02-8A3), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | TRIM56 |
|---|---|
| Clone | S02-8A3 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696053
|
Novus Biologicals
NBP3-19795-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618203
|
Novus Biologicals
NBP3-19795-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM56 Monoclonal antibody specifically detects TRIM56 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| TRIM56 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp667O116, E3 ubiquitin-protein ligase TRIM56, EC 6.3.2.-, RING finger protein 109, RNF109FLJ35608, tripartite motif containing 56, tripartite motif-containing 56, Tripartite motif-containing protein 56 | |
| A synthetic peptide of human TRIM56 (Uniprot # Q9BRZ2) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S02-8A3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 81844 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title