missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM58 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81870FR
This item is not returnable.
View return policy
Description
TRIM58 Polyclonal antibody specifically detects TRIM58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| TRIM58 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 680 | |
| BIA2, DKFZp434C091, Protein BIA2, tripartite motif containing 58, tripartite motif-containing 58, tripartite motif-containing protein 58 | |
| TRIM58 antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TRIM58. Amino Acid Squence: DTLPRKGETTPSPENGV | |
| 0.1 mL | |
| Zinc Finger | |
| 25893 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction