missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM66 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555607
This item is not returnable.
View return policy
Description
Immunogen sequence: RLERLKICAA SSGEMPVFKL KPQKNDQDGS FLLIIECGTE SSSMSIKVSQ DRLSEATQAP GLEGRKVTVT SLAGQRPPEV EGTSPEEHRL IPRTP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
TRIM66 gene ontology annotations related to this gene include zinc ion binding.
Specifications
| TRIM66 | |
| Polyclonal | |
| Unconjugated | |
| TRIM66 | |
| C11orf29; KIAA0298; Kiaa0298-hp; Tif1d; TIF1DELTA; transcriptional intermediary factor 1 delta; Trim66; tripartite motif containing 66; tripartite motif-containing 66; tripartite motif-containing protein 66 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9866 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15016 | |
| TRIM66 | |
| Recombinant protein corresponding to Human TRIM66. Recombinant protein control fragment (Product #RP-93610). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction