missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM72 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22151-1-AP-150UL
This item is not returnable.
View return policy
Description
Muscle-specific protein that plays a central role in cell membrane repair by nucleating the assembly of the repair machinery at injury sites. Specifically binds phosphatidylserine. Acts as a sensor of oxidation: upon membrane damage, entry of extracellular oxidative environment results in disulfide bond formation and homooligomerization at the injury site. This oligomerization acts as a nucleation site for recruitment of TRIM72-containing vesicles to the injury site, leading to membrane patch formation. Probably acts upstream of the Ca(2+)-dependent membrane resealing process. Required for transport of DYSF to sites of cell injury during repair patch formation. Regulates membrane budding and exocytosis. May be involved in the regulation of the mobility of KCNB1-containing endocytic vesicles.Specifications
| TRIM72 | |
| Polyclonal | |
| Unconjugated | |
| TRIM72 | |
| hMG53, Mg53, Mitsugumin 53, TRIM72, tripartite motif containing 72 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 434246, 493829 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q1XH17, Q6ZMU5 | |
| Trim72 | |
| TRIM72 Fusion Protein Ag17756 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction