Learn More
TRIM9 Monoclonal Antibody (OTI1G10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800045S
Encompass_Preferred
 View more versions of this product
Product Code. 17527432
  • € 242.00 / 30µL
Description
TRIM9 Monoclonal antibody specifically detects TRIM9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
Specifications
TRIM9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9C026
Mouse
Affinity chromatography
RUO
114088
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Flow Cytometry, Immunocytochemistry, Western Blot
OTI1G10
Unconjugated
TRIM9
E3 ubiquitin-protein ligase TRIM9; fj48c11; homolog of rat RING finger Spring; hypothetical protein LOC336099; Kiaa0282; RING finger protein 91; RING-type E3 ubiquitin transferase TRIM9; RNF91; SNAP-25-interacting RING finger protein; Spring; Trim9; tripartite motif containing 9; tripartite motif protein 9; tripartite motif-containing 9; tripartite motif-containing protein 9; tripartite motif-containing protein 9; E3 ubiquitin-protein ligase TRIM9; Unknown (protein for MGC:137652); wu:fj48c11; zgc:77357
Human recombinant protein fragment corresponding to amino acids 284-669 of human TRIM9 produced in E.coli.