missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM9 Monoclonal Antibody (OTI3C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800062S
This item is not returnable.
View return policy
Description
TRIM9 Monoclonal antibody specifically detects TRIM9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| TRIM9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q93008 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8239 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI3C9 | |
| Unconjugated | |
| USP9X | |
| E3 ubiquitin-protein ligase TRIM9; fj48c11; homolog of rat RING finger Spring; hypothetical protein LOC336099; Kiaa0282; RING finger protein 91; RING-type E3 ubiquitin transferase TRIM9; RNF91; SNAP-25-interacting RING finger protein; Spring; Trim9; tripartite motif containing 9; tripartite motif protein 9; tripartite motif-containing 9; tripartite motif-containing protein 9; tripartite motif-containing protein 9; E3 ubiquitin-protein ligase TRIM9; Unknown (protein for MGC:137652); wu:fj48c11; zgc:77357 | |
| Human recombinant protein fragment corresponding to amino acids 2246-2570 of human USP9X produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction