missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM9 Monoclonal Antibody (OTI5G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800067
This item is not returnable.
View return policy
Description
TRIM9 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.Specifications
| TRIM9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9C026 | |
| TRIM9 | |
| Human recombit protein fragment corresponding to amino acids 284-669 of human TRIM9 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5G5 | |
| Unconjugated | |
| TRIM9 | |
| RNF91, SPRING | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 114088 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction