Learn More
Trimeprazine Tartrate 1.0 mg/ml in Methanol (as free base), LoGiCal
Product Code. 30461268
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461268 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461268 Supplier LoGiCal Supplier No. LGCAMP0958.0001
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Alimemazine tartrate
CAS 4330-99-8
Analyte or Component Names Alimemazine tartrate
Shipping Condition Room Temperature
Molecular Formula 2 C18 H22 N2 S . C4 H6 O6
Certifications/Compliance ISO 17034
Synonym Phenothiazine, 10-[3-(dimethylamino)-2-methylpropyl]-, tartrate (2:1) (8CI), 10-(3-Dimethylamino-2-methylpropyl)phenothiazine tartrate, Alimemazine tartrate, NSC 17475, Panectyl, Repeltin, Temaril, Temaril 16, Theralene, Trimeprazine Tartrate, Vallergan, Vanectyl, 10H-Phenothiazine-10-propanamine, N,N,beta-trimethyl-, (2R,3R)-2,3-dihydroxybutanedioate (2:1), 10H-Phenothiazine-10-propanamine, N,N,beta-trimethyl-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1)
InChI Formula InChI=1S/2C18H22N2S.C4H6O6/c2*1-14(12-19(2)3)13-20-15-8-4-6-10-17(15)21-18-11-7-5-9-16(18)20;5-1(3(7)8)2(6)4(9)10/h2*4-11,14H,12-13H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1
Solution Type Single Solution
SMILES CC(CN(C)C)CN1c2ccccc2Sc3ccccc13.CC(CN(C)C)CN4c5ccccc5Sc6ccccc46.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 746.98
Quantity 1 mL
Formula Weight 746.3172 g/mol