Learn More
Trimeprazine Tartrate, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Trimeprazine Tartrate
Specifications
Specifications
| Chemical Name or Material | Alimemazine tartrate |
| CAS | 4330-99-8 |
| Analyte or Component Names | Alimemazine tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C18 H22 N2 S . C4 H6 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Phenothiazine, 10-[3-(dimethylamino)-2-methylpropyl]-, tartrate (2:1) (8CI), 10-(3-Dimethylamino-2-methylpropyl)phenothiazine tartrate, Alimemazine tartrate, NSC 17475, Panectyl, Repeltin, Temaril, Temaril 16, Theralene, Trimeprazine Tartrate, Vallergan, Vanectyl, 10H-Phenothiazine-10-propanamine, N,N,beta-trimethyl-, (2R,3R)-2,3-dihydroxybutanedioate (2:1), 10H-Phenothiazine-10-propanamine, N,N,beta-trimethyl-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1) |
| InChI Formula | InChI=1S/2C18H22N2S.C4H6O6/c2*1-14(12-19(2)3)13-20-15-8-4-6-10-17(15)21-18-11-7-5-9-16(18)20;5-1(3(7)8)2(6)4(9)10/h2*4-11,14H,12-13H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1 |
| SMILES | CC(CN(C)C)CN1c2ccccc2Sc3ccccc13.CC(CN(C)C)CN4c5ccccc5Sc6ccccc46.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine |
| Show More |