Learn More
Trimethobenzamide Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Trimethobenzamide Hydrochloride |
| CAS | 554-92-7 |
| Analyte or Component Names | Trimethobenzamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 N2 O5 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzamide, N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4,5-trimethoxy-, hydrochloride (1:1), Benzamide, N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4,5-trimethoxy-, monohydrochloride (9CI), Benzamide, N-[p-[2-(dimethylamino)ethoxy]benzyl]-3,4,5-trimethoxy-, hydrochloride (6CI,7CI), Benzamide, N-[p-[2-(dimethylamino)ethoxy]benzyl]-3,4,5-trimethoxy-, monohydrochloride (8CI), 4-(2-Dimethylaminoethoxy)-N-(3,4,5-trimethoxybenzoyl)benzylamine hydrochloride, Anaus, N-[p-[2-(Dimethylamino)ethoxy]benzyl]-3,4,5-trimethoxybenzamide hydrochloride, Ro 2-9578, Tigan, Tigan hydrochloride, Trimethobenzamide hydrochloride, Xametina |
| InChI Formula | InChI=1S/C21H28N2O5.ClH/c1-23(2)10-11-28-17-8-6-15(7-9-17)14-22-21(24)16-12-18(25-3)20(27-5)19(13-16)26-4;/h6-9,12-13H,10-11,14H2,1-5H3,(H,22,24);1H |
| Solution Type | Neat |
| SMILES | Cl.COc1cc(cc(OC)c1OC)C(=O)NCc2ccc(OCCN(C)C)cc2 |
| Show More |