missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim, 99.8%, MP Biomedicals™

Product Code. 11415492 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11415492

Brand: MP Biomedicals 0219552780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.323
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
Synonym 2,4-Diamino-5-[3,4,5-trimethoxy-benzyl]pyrimidine
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N

Specifications

Type Trimethoprim
Physical Form Powder
Color White
Quantity 100 g
Assay Percent Range 0.998
Formula Weight 290.3
Chemical Name or Material Trimethoprim
compliance-icons
Product Identifier
  • Trimethoprim
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Trimethoprim
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.