Learn More
Trimethyl(prop-2-yn-1-yloxy)silane, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 5582-62-7 |
| Molecular Formula | C6H12OSi |
| MDL Number | MFCD00042844 |
| Quantity | 25 g |
| SMILES | C#CCO[Si](C)(C)C |
| Molecular Weight (g/mol) | 128.24 |
| Percent Purity | 97% |
| Chemical Name or Material | Trimethyl(prop-2-yn-1-yloxy)silane |