missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIML1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113158
This item is not returnable.
View return policy
Description
TRIML1 Polyclonal Antibody for Western Blot, ELISA
Probable E3 ubiquitin-protein ligase which plays an important role in blastocyst development.
Specifications
| TRIML1 | |
| Polyclonal | |
| Unconjugated | |
| Triml1 | |
| 4933403D14; BC050188; probable E3 ubiquitin-protein ligase TRIML1; RGD1305337; RING finger protein 209; RING-type E3 ubiquitin transferase TRIML1; RNF209; Triml1; tripartite motif family like 1; tripartite motif family-like 1; tripartite motif family-like protein 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 244448, 339976 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BVP1, Q8N9V2 | |
| Triml1 | |
| Recombinant Human Probable E3 ubiquitin-protein ligase TRIML1 protein (277-468AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction