missing translation for 'onlineSavingsMsg'
Learn More

Trioctylsilane, 95%, ACROS Organics™

Brand:  Acros Organics 434900250

Additional Details : CAS : 18765-09-8 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 11995941

  • € 185.00 / 25g


Estimated Shipment: 16-08-2021
Add to basket

Description

Description

Reduction reagent
Specifications

Chemical Identifiers

18765-09-8
368.76
trioctylsilane, silane, trioctyl, tri-n-octylsilane, trioctylsilyl
trioctylsilicon
C24H52Si
QTKHQYWRGFZFHG-UHFFFAOYSA-N
6327367
CCCCCCCC[Si](CCCCCCCC)CCCCCCCC

Specifications

Trioctylsilane
113°C
Glass bottle
163.0°C to 165.0°C (0.2mmHg)
25g
94% min. (GC)
trioctylsilane, silane, trioctyl, tri-n-octylsilane, trioctylsilyl
CCCCCCCC[Si](CCCCCCCC)CCCCCCCC
368.76
368.76
95%
0.8210g/mL
Authentic
0.821
Colorless
18765-09-8
C24H52Si
QTKHQYWRGFZFHG-UHFFFAOYSA-N
trioctylsilicon
6327367
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Trioctylsilane, 95%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates