missing translation for 'onlineSavingsMsg'
Learn More

Trioctylsilane, 95%, Thermo Scientific Chemicals

Catalog Number p-4228948
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
18765-09-8
C24H52Si
368.76
QTKHQYWRGFZFHG-UHFFFAOYSA-N
trioctylsilane, silane, trioctyl, tri-n-octylsilane, trioctylsilyl
6327367
trioctylsilicon
CCCCCCCC[Si](CCCCCCCC)CCCCCCCC
This item is not returnable. View return policy
18765-09-8
C24H52Si
368.76
QTKHQYWRGFZFHG-UHFFFAOYSA-N
trioctylsilane, silane, trioctyl, tri-n-octylsilane, trioctylsilyl
6327367
trioctylsilicon
CCCCCCCC[Si](CCCCCCCC)CCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8210g/mL
Boiling Point 163.0°C to 165.0°C (0.2 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.821
Formula Weight 368.76
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Trioctylsilane
Show More Show Less
compliance-icons
Product Identifier
  • Trioctylsilane, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'