Learn More
Trioctyltin chloride 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30322437
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322437 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30322437 Supplier Dr. Ehrenstorfer Supplier No. DREV17892000ME1000
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mL
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol