missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA572919

Product Code. 15931575

  • € 530.00 / 0.05mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRIP Polyclonal Antibody for Western Blot, IHC (P), IP

TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRIP
Polyclonal
Unconjugated
TRAIP
E3 ubiquitin-protein ligase TRAIP; RING finger protein 206; RING-type E3 ubiquitin transferase TRAIP; RNF206; SCKL9; TRAF interacting protein; TRAF-interacting protein; TRAIP; Trip
Rabbit
50 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conc. Not Determined
Whole serum with 0.05% sodium azide
Q9BWF2
TRAIP
A synthetic peptide corresponding to amino acids 294-313 of human TRIP.
RUO
10293
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.