missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111072
This item is not returnable.
View return policy
Description
Immunogen sequence: LNLPPVASET VDRLVLESPA PVEVNLKLRR PSFRDDIDLN ATFDVDTPPA RPSSSQHGYY EKLCLEKSHS PIQDVPKKIC KGPRKESQLS L Highest antigen sequence indentity to the following orthologs: Rat - 65%, Mouse - 60%.
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.
Specifications
| TRIP | |
| Polyclonal | |
| Unconjugated | |
| TRAIP | |
| E3 ubiquitin-protein ligase TRAIP; RING finger protein 206; RING-type E3 ubiquitin transferase TRAIP; RNF206; SCKL9; TRAF interacting protein; TRAF-interacting protein; TRAIP; Trip | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10293 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BWF2 | |
| TRAIP | |
| Recombinant Protein corresonding to Human TRIP. Recombinant protein control fragment (Product #RP-104300). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction