Learn More
Invitrogen™ TRIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143982
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Daudi whole cell, human U251 whole cell, rat brain tissue, mouse brain tissue. ICC/IF: Hela cell. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.
Specifications
| TRIP | |
| Polyclonal | |
| Unconjugated | |
| TRAIP | |
| E3 ubiquitin-protein ligase TRAIP; RING finger protein 206; RING-type E3 ubiquitin transferase TRAIP; RNF206; SCKL9; TRAF interacting protein; TRAF-interacting protein; TRAIP; Trip | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10293, 22036, 367167 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q8VIG6, Q9BWF2 | |
| TRAIP | |
| E.coli-derived human TRIP/TRAIP recombinant protein (Position: M1-S469). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.