Learn More
TRIP6 Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human TRIP6.
Brand: Abnova PAB29450.100uL
Description
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]
Sequence: RPGSLDAEIDLLSSTLAELNGGRGHASRRPDRQAYEPPPPPAYRTGSLKPNPASPSpecifications
| TRIP6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant human TRIP6. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| TRIP6 | |
| Recombinant protein corresponding to human TRIP6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| MGC10556/MGC10558/MGC29959/MGC3837/MGC4423/OIP1/ZRP-1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7205 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.