missing translation for 'onlineSavingsMsg'
Learn More

Tripeptidyl peptidase II Antibody (4A3) [DyLight 755], Novus Biologicals Biologicals™

Product Code. 30522418 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522418 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522418 Supplier Novus Biologicals Supplier No. NBP343048IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tripeptidyl peptidase II Monoclonal antibody specifically detects Tripeptidyl peptidase II in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tripeptidyl peptidase II
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 4A3
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.14.10, FLJ40359, TPP-2, TPP-II, Tripeptidyl aminopeptidase, tripeptidyl peptidase II, tripeptidyl-peptidase 2, Tripeptidyl-peptidase II
Host Species Mouse
Immunogen tripeptidyl peptidase II - (Uniprot# P29144)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 7174
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.