missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride, 98%, Thermo Scientific™

Catalog Number p-4228952
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
99662-46-1
C24H21ClNP·ClH
426.32
JHQWTRCLYUFMSJ-UHFFFAOYSA-M
triphenyl 2-pyridylmethyl phosphonium chloride hydrochloride, triphenyl pyridin-2-ylmethyl phosphanium hydrochloride chloride, acmc-20ajcr, triphenyl 2-pyridylmethyl-phosphonium chloride hydrochloride, triphenyl 2-pyridylmethyl phosphonium ch loride hydrochloride, triphenyl pyridin-2-yl methyl phosphanium chloride-hydrogen chloride 1/1/1
16213046
triphenyl(pyridin-2-ylmethyl)phosphanium;chloride;hydrochloride
C1=CC=C(C=C1)[P+](CC2=CC=CC=N2)(C3=CC=CC=C3)C4=CC=CC=C4.Cl.[Cl-]
This item is not returnable. View return policy
99662-46-1
C24H21ClNP·ClH
426.32
JHQWTRCLYUFMSJ-UHFFFAOYSA-M
triphenyl 2-pyridylmethyl phosphonium chloride hydrochloride, triphenyl pyridin-2-ylmethyl phosphanium hydrochloride chloride, acmc-20ajcr, triphenyl 2-pyridylmethyl-phosphonium chloride hydrochloride, triphenyl 2-pyridylmethyl phosphonium ch loride hydrochloride, triphenyl pyridin-2-yl methyl phosphanium chloride-hydrogen chloride 1/1/1
16213046
triphenyl(pyridin-2-ylmethyl)phosphanium;chloride;hydrochloride
C1=CC=C(C=C1)[P+](CC2=CC=CC=N2)(C3=CC=CC=C3)C4=CC=CC=C4.Cl.[Cl-]
Melting Point 241.0°C to 244.5°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. On dry basis (Argentometry)
Quantity 5 g
Formula Weight 426.32
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride
compliance-icons
Product Identifier
  • Triphenyl(2-pyridylmethyl)phosphonium chloride hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'