Learn More
Tris(2,4-di-tert-butylphenyl)phosphate, Mikromol
Description
Tris(2,4-di-tert-butylphenyl)phosphate, Mikromol
Specifications
Specifications
| Chemical Name or Material | Tris(2,4-di-tert-butylphenyl) phosphate |
| CAS | 95906-11-9 |
| Analyte or Component Names | Tris(2,4-di-tert-butylphenyl) phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C42 H63 O4 P |
| Certifications/Compliance | ISO 9001 |
| Synonym | Phenol, 2,4-bis(1,1-dimethylethyl)-, 1,1',1''-phosphate, Phenol, 2,4-bis(1,1-dimethylethyl)-, phosphate (3:1) (9CI), Alcanox 240, D 16-834, P 3, Tris(2,4-di-tert-butylphenyl) phosphate, Tris(2,4-di-tert-butylphenyl) phosphate |
| InChI Formula | InChI=1S/C42H63O4P/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)44-47(43,45-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)46-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18/h19-27H,1-18H3 |
| Solution Type | Neat |
| SMILES | CC(C)(C)c1ccc(OP(=O)(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3ccc(cc3C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C |
| Show More |