missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-di-tert-butylphenyl)phosphate, Tris(2,4-di-tert-butylphenyl)phosphate, TRC

Product Code. 17948884
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17948884 100 mg 100mg
17948494 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17948884 Supplier Toronto Research Chemicals Supplier No. TRCT884500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C42 H63 O4 P
Molecular Weight (g/mol) 662.92
Synonym Phenol, 2,4-bis(1,1-dimethylethyl)-, 1,1',1' '-phosphate, Phenol, 2,4-bis(1,1-dimethylethyl)-, phosphate (3:1) (9CI), Alcanox 240, D 16-834, P 3, Tris(2,4-di-tert-butylphenyl) phosphate
IUPAC Name tris(2,4-ditert-butylphenyl) phosphate
SMILES CC(C)(C)c1ccc(OP(=O)(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)Oc3ccc(cc3C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C

Specifications

CAS 95906-11-9
Quantity 100 mg
InChI Formula InChI=1S/C42H63O4P/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)44-47(43,45-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)46-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18/h19-27H,1-18H3
Formula Weight 662.4464
Percent Purity >95%
Chemical Name or Material Tris(2,4-di-tert-butylphenyl) phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.